1-Butanol ≥99.4% (by GC, corrected for water content), AR® (ACS) ACS, Macron Fine Chemicals™
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
|
Warning
|
Synonyms:
Propylcarbinol, Methylolpropane, Butan-1-ol, Butyl alcohol, n-Butyl alcohol, Butyl hydroxide, 1-Butyl alcohol, Butyl Alcohol, 1-Hydroxybutane, 1-Hydroxy butane, n-Butan-1-ol, Propyl carbinol, n-Butanol, n-Butyl Alcohol
Management of Change (MOC) category = N
3000-26
3000-04
3000-08
MK300026EA
23938.44
USD
MK300026
MK300004
MK300008
1-Butanol ≥99.4% (by GC, corrected for water content), AR® (ACS) ACS, Macron Fine Chemicals™
1-Butanol
AR—The standard Macron Fine Chemicals™ grade of analytical reagents; suitable for laboratory and general use. If the reagent also meets the requirements of the American Chemical Society Committee on Analytical Reagents, it will be denoted as an AR (ACS) r
Formula:
H₃C(CH₂)₂CH₂OH MW: 74.12 g/mol Boiling Pt: 117.6 Melting Pt: -89 Density: 0.812 g/cm³ (20 °C) Flash Pt: 37 °C (closed cup) Storage Temperature: Ambient |
MDL Number:
MFCD00002964 CAS Number: 71-36-3 EINECS: 200-751-6 UN: 1120 ADR: 3 III REACH: 01-2119484630-38 Merck Index: 13,01539 |
|
Specification Test Results
Meets ACS Reagent Chemical Requirements | |
For Laboratory,Research,or Manufacturing Use | |
ACS - Assay (CH₃CH₂CH₂CH₂OH) (by GC, corrected for water) | ≥99.4 % |
ACS - Butyl Ether (C₈H₁₈O)(by GC) | ≤0.2 % |
ACS - Carbonyl Compounds (as Butyraldehyde) | ≤0.01 % |
ACS - Color (APHA) | ≤10 |
ACS - Residue after Evaporation | ≤0.005 % |
ACS - Titrable Acid (as CH₃COOH) (meq/g) | ≤0.0008 |
ACS - Water (H₂O)(by Karl Fischer titrn) | ≤0.1 % |
Appearance (clear, colorless liquid) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...