Iodine 0.10 N (N/10) in aqueous solution, StandARd® volumetric solution, Macron Fine Chemicals™

Supplier: AVANTOR PERF MAT - MACRON LAB
Danger


Management of Change (MOC) category = NI

2962-60 2962-07 MACR2962-07
MK296260CS 695.41 USD
MK296260 JT2962-07 MK2962-07
Iodine 0.10 N (N/10) in aqueous solution, StandARd® volumetric solution, Macron Fine Chemicals™
Iodine
Formula: I₂
MW: 253.81 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011355
CAS Number: 7553-56-2
EINECS: 231-442-4
UN: 3264
ADR: 8,II

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Normality 0.0995 - 0.1005
Appearance (dark brown to red-brown solution)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR