Hydrochloric acid 36.5% - 38.0% NF, FCC, ACS, Macron Fine Chemicals™
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
|
Danger
|
Synonyms:
Hydrochloric acid solution, Muriatic acid
Management of Change (MOC) category = R
2612-44
2612-14
2612-19
2612-24
2612-06
MACR2612-46
MK261244CS
317.89
USD
MK261244
MK2612-14
MK261219
MK261224
MK261206
MK2612-46
Hydrochloric acid 36.5% - 38.0% NF, FCC, ACS, Macron Fine Chemicals™
Hydrochloric acid
Formula:
HCl MW: 36.46 g/mol Boiling Pt: 110 °C (1013 hPa) Melting Pt: –30 °C Density: 1.18 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011324 CAS Number: 7647-01-0 EINECS: 231-595-7 UN: 1789 ADR: 8,II |
Specification Test Results
GMP Manufactured Product | |
Meets ACS Reagent Chemical Requirements | |
Meets F.C.C. Requirements | |
Meets N.F. Requirements | |
Food GMP Manufactured Product | |
Store below 25°C | |
CAUTION: For Manufacturing, processing or repackaging | |
Bulk Food Chemical | |
Bulk Pharmaceutical Chemical | |
ACS - Assay (HCl) (ACS) | 36.5 - 38.0 % |
ACS - Ammonium (NH₄) | ≤ 3 ppm |
ACS - Arsenic (As) | ≤ 0.010 ppm |
ACS - Bromide (Br) | ≤ 0.005 % |
ACS - Color (APHA) | ≤ 10 |
ACS - Extractable Organic Substances | ≤ 5 ppm |
ACS - Free Chlorine (as Cl) | ≤ 1 ppm |
ACS - Residue after Ignition | ≤ 5 ppm |
ACS - Sulfate (SO₄) | ≤ 1 ppm |
ACS - Sulfite (SO₃) | ≤ 1 ppm |
ACS - Heavy Metals (as Pb) | ≤ 1 ppm |
Trace Impurities - ACS - Iron (Fe) | ≤ 0.200 ppm |
NF - Assay (HCl)(NF) | 36.5 - 38.0 % |
NF - Bromide or Iodide | Passes Test |
NF - Free Bromine or Chlorine | Passes Test |
NF - Identification | Passes Test |
NF - Residue on Ignition | ≤ 0.008 % |
NF - Sulfate (NF)(SO₄) | Passes Test |
NF - Sulfite (NF)(SO₃) | Passes Test |
FCC - Assay (HCl)(FCC) | 36.5 - 38.0 % |
FCC - Baume Degrees | 22.7 - 23.4 |
FCC - Color (APHA) | Passes Test |
FCC - Identification | Passes Test |
FCC - Iron (Fe) | ≤ 0.2 ppm |
FCC - Lead (Pb) (mg/kg) | ≤ 1 |
FCC - Mercury (Hg)(mg/kg) | ≤ 0.10 |
FCC - Nonvolatile Residue | ≤ 0.5 % |
Total Organic Compounds - Non-Fluorine Containing (mg/kg) | ≤ 5 |
Total Organic Compounds - Benzene (mg/kg) | ≤ 0.05 |
Total Organic Compounds - Fluorinated Organic Compounds(mg/kg) | ≤ 25 |
Total Organic Compounds - Oxidizing Substances | ≤ 0.003 % |
Reducing Substances | ≤ 0.007 % |
Specific Gravity | 1.1856 - 1.1924 |
Sulfate (SO₄) | ≤ 0.5 % |
Appearance | Passes Test |
No Class 1,2,3 or other solvents are used or produced in the | |
manufacturing or purification of the product. | |
Elemental Impurities (USP 232, EP 5.20) - Information on elemental | |
impurities for this product is available on the associated Product | |
Regulatory Data Sheet and elemental impurity profile report. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...