EDTA (ethylenediamine tetraacetic acid) 99.4-100.6%, powder, AR® ACS, Macron Fine Chemicals™
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
|
Warning
|
Management of Change (MOC) category = N
2580-12
MK258012CS
1406.24
USD
MK258012
EDTA (ethylenediamine tetraacetic acid) 99.4-100.6%, powder, AR® ACS, Macron Fine Chemicals™
EDTA (Ethylenediamine tetraacetic acid)
Powder.
Formula:
C₁₀H₁₆N₂O₈ Boiling Pt: 614 °C (1013 hPa) Melting Pt: 220 °C Density: 0.86 g/cm³ (20 °C) Flash Pt: >100 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003541 CAS Number: 60-00-4 EINECS: 200-449-4 REACH: 01-2119486399-18 Merck Index: 13,03546 |
|
Specification Test Results
Meets ACS Reagent Chemical Requirements | |
For Laboratory,Research,or Manufacturing Use | |
ACS - Assay (C₁₀H₁₆N₂O₈) | 99.4 - 100.6 % |
ACS - Calcium (Ca) | ≤0.001 % |
ACS - Heavy Metals (as Pb) | ≤0.001 % |
ACS - Insoluble in Dilute NH₄OH | ≤0.005 % |
ACS - Iron (Fe) | ≤0.005 % |
ACS - Magnesium (Mg) | ≤5 ppm |
ACS - Nitriloacetic Acid | ≤0.1 % |
ACS - Residue after Ignition | ≤0.2 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...