Acetic acid 36% NF, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger


Management of Change (MOC) category = R

2488-44
MK248844CS 2153.32 USD
MK248844
Acetic acid 36% NF, Macron Fine Chemicals™
Acetic acid
Formula: H₃CCOOH
Storage Temperature: Ambient
MDL Number: MFCD00036152
CAS Number: 64-19-7
EINECS: 200-580-7
UN: 2790
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4ecdb21d8177dd197c843d4b5187410fee102541&vid=6c26d1716b942e60cb8b3ec0f91e6623fed5b3db&prid=web&clid=SAPDAM

Order Now

Specification Test Results

GMP Manufactured Product
Meets N.F. Requirements
CAUTION: For Manufacturing, processing or repackaging
Bulk Pharmaceutical Chemical
NF - Assay (CH₃COOH) 36.0 - 37.0 %
NF - Chloride (Cl) Passes Test
NF - Identification Passes Test
NF - Nonvolatile Residue ≤0.005 %
NF - Readily Oxidizable Substances Passes Test
NF - Sulfate (SO₄) Passes Test
Appearance (clear, colorless liquid)
Preserve in tight containers, and store at room temperature.
Residual Solvents: Only the Class 3 Solvents acetone, isopropyl acetate,
and propyl acetate are expected to be present. Residual concentration of all Class 3 Solvents combined is <0.1%.
Elemental Impurities (USP 232, EP 5.20) - Information on elemental
impurities for this product is available on the associated Product
Regulatory Data Sheet and elemental impurity profile report.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR