Acetone ≥99.5%, AR® ACS, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger


Management of Change (MOC) category = N

2443-10
MK244310CS 1385.72 USD
MK244310
Acetone ≥99.5%, AR® ACS, Macron Fine Chemicals™
Acetone
Formula: CH₃COCH₃
Boiling Pt: 56.2 °C (1013 hPa)
Melting Pt: –95.4 °C
Density: 0.792 g/cm³ (20 °C)
Flash Pt: <–20 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00008765
CAS Number: 67-64-1
EINECS: 200-662-2
UN: 1090
ADR: 3,II
REACH: 01-2119471330-49
Merck Index: 13,00067
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3de5d8e3bf7a94692d684c8f0ccdfb8500fde76d&vid=24fb2e0ec86974b1803780c7d5b23804a085df64&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
ACS - Assay (CH₃COCH₃) (by GC, corrected for water) ≥99.5 %
ACS - Aldehydes (as HCHO) ≤0.002 %
ACS - Color (APHA) ≤10
ACS - Isopropyl Alcohol (CH₃CHOHCH₃)(by GC) ≤0.05 %
ACS - Methanol (CH₃OH)(by GC) ≤0.05 %
ACS - Residue after Evaporation ≤0.001 %
ACS - Solubility in H₂O Passes Test
ACS - Substances Reducing Permanganate Passes Test
ACS - Titrable Acid (meq/g) ≤0.0003
ACS - Titrable Base (meq/g) ≤0.0006
ACS - Water (H₂O) ≤0.5 %
Appearance (clear, colorless liquid)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR