Acetone ≥99.5% (by GC, corrected for water content), AR® ACS for histology, Macron Fine Chemicals™
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
|
Danger
|
Synonyms:
2-Propanone, DMK, Propanone, Dimethyl ketone
Management of Change (MOC) category = N
2440-02
2440-22
2440-06
2440-10
2440-16
2440-24
MK244002CS
830.02
USD
MK244002
MK244022
MK244006
MK2440-10
MK244016
MK244024
Acetone ≥99.5% (by GC, corrected for water content), AR® ACS for histology, Macron Fine Chemicals™
Acetone
For histological use.
Formula:
CH₃COCH₃ Boiling Pt: 56.2 °C (1013 hPa) Melting Pt: –95.4 °C Density: 0.792 g/cm³ (20 °C) Flash Pt: <–20 °C (closed cup) Storage Temperature: Ambient |
MDL Number:
MFCD00008765 CAS Number: 67-64-1 EINECS: 200-662-2 UN: 1090 ADR: 3,II REACH: 01-2119471330-49 Merck Index: 13,00067 |
|
Specification Test Results
Meets ACS Reagent Chemical Requirements | |
For Laboratory,Research,or Manufacturing Use | |
ACS - Assay (CH₃COCH₃) (by GC, corrected for water) | ≥99.5 % |
ACS - Aldehydes (as HCHO) | ≤0.002 % |
ACS - Color (APHA) | ≤10 |
ACS - Isopropyl Alcohol (CH₃CHOHCH₃)(by GC) | ≤0.05 % |
ACS - Methanol (CH₃OH)(by GC) | ≤0.05 % |
ACS - Residue after Evaporation | ≤0.001 % |
ACS - Solubility in H₂O | Passes Test |
ACS - Substances Reducing Permanganate | Passes Test |
ACS - Titrable Acid (meq/g) | ≤0.0003 |
ACS - Titrable Base (meq/g) | ≤0.0006 |
ACS - Water (H₂O) | ≤0.5 % |
Appearance (clear, colorless liquid) | |
Date Received: / / | |
Date Opened: / / |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...