Acetone ≥99.5% (by GC, corrected for water content), AR® ACS for histology, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger

Synonyms: 2-Propanone, DMK, Propanone, Dimethyl ketone

Management of Change (MOC) category = N

2440-02 2440-22 2440-06 2440-10 2440-16 2440-24
MK244002CS 830.02 USD
MK244002 MK244022 MK244006 MK2440-10 MK244016 MK244024
Acetone ≥99.5% (by GC, corrected for water content), AR® ACS for histology, Macron Fine Chemicals™
Acetone

For histological use.

Formula: CH₃COCH₃
MW: 58.08 g/mol
Boiling Pt: 56.2
Melting Pt: -95.4
Density: 0.792 g/cm³ (20 °C)
Flash Pt: <–20 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00008765
CAS Number: 67-64-1
EINECS: 200-662-2
UN: 1090
ADR: 3 II
REACH: 01-2119471330-49
Merck Index: 13,00067
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3de5d8e3bf7a94692d684c8f0ccdfb8500fde76d&vid=24fb2e0ec86974b1803780c7d5b23804a085df64&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
ACS - Assay (CH₃COCH₃) (by GC, corrected for water) ≥99.5 %
ACS - Aldehydes (as HCHO) ≤0.002 %
ACS - Color (APHA) ≤10
ACS - Isopropyl Alcohol (CH₃CHOHCH₃)(by GC) ≤0.05 %
ACS - Methanol (CH₃OH)(by GC) ≤0.05 %
ACS - Residue after Evaporation ≤0.001 %
ACS - Solubility in H₂O Passes Test
ACS - Substances Reducing Permanganate Passes Test
ACS - Titrable Acid (meq/g) ≤0.0003
ACS - Titrable Base (meq/g) ≤0.0006
ACS - Water (H₂O) ≤0.5 %
Appearance (clear, colorless liquid)
Date Received: / /
Date Opened: / /


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR