L(+)-Tartaric acid ≥99%, granules, AR® ACS, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger

Synonyms: L-Tartaric Acid, (2R,3R)-(+)-Tartaric acid, Tartaric Acid, (R,R)-(+)-2,3-Dihydroxysuccinic acid, (R,R)-(+)-Tartaric acid, (2R,3R)-2,3-Dihydroxysuccinic acid

Management of Change (MOC) category = N

2312-04 2312-06
MK231204CS 3113.92 USD
MK231204 MK231206
L(+)-Tartaric acid ≥99%, granules, AR® ACS, Macron Fine Chemicals™
L(+)-Tartaric acid

Granular.

Formula: C₄H₆O₆
MW: 150.09 g/mol
Boiling Pt: 399
Melting Pt: 168…170 °C
Density: 1.759 g/cm³ (20 °C)
Flash Pt: 150 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00064207
CAS Number: 87-69-4
EINECS: 201-766-0
REACH: 01-2119537204-47
Merck Index: 13,09158
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d4e859e52081482a351d52322717614aad450742&vid=879c292903ddee7a59d5d6b2d969c4c8fc6eac75&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
ACS - Assay ≥99.0 %
ACS - Chloride (Cl) ≤0.001 %
ACS - Heavy Metals (as Pb) ≤5 ppm
ACS - Insoluble Matter ≤0.005 %
ACS - Iron (Fe) ≤5 ppm
ACS - Oxalate (C₂O₄) Passes Test
ACS - Phosphate (PO₄) ≤0.001 %
ACS - Residue after Ignition ≤0.02 %
ACS - Sulfur Compounds (as S) ≤0.002 %
Appearance (fine, colorless crystalline granules)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR