L(+)-Tartaric acid 99.7-100.5% (dried basis), fine-granular NF, FCC, Macron Fine Chemicals™
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
|
Danger
|
Synonyms:
L-Tartaric Acid, (2R,3R)-(+)-Tartaric acid, Tartaric Acid, (R,R)-(+)-2,3-Dihydroxysuccinic acid, (R,R)-(+)-Tartaric acid, (2R,3R)-2,3-Dihydroxysuccinic acid
Management of Change (MOC) category = R
2307-04
2307-25
2307-QM
MK230704CS
2471.12
USD
MK230704
JT2307-25
JT2307-QM
L(+)-Tartaric acid 99.7-100.5% (dried basis), fine-granular NF, FCC, Macron Fine Chemicals™
L(+)-Tartaric acid
White crystalline granules. Actual signed Certificate of Lot Analysis provided.
Formula:
C₄H₆O₆ MW: 150.09 g/mol Boiling Pt: 399 °C (1013 hPa) Melting Pt: 206 °C Density: 1.759 g/cm³ (20 °C) Flash Pt: > 100 °C (closed cup) Storage Temperature: Ambient |
MDL Number:
MFCD00064207 CAS Number: 87-69-4 EINECS: 201-766-0 REACH: 01-2119537204-47 Merck Index: 13,09158 |
|
Specification Test Results
GMP Manufactured Product | |
Meets N.F. Requirements | |
Meets F.C.C. Requirements | |
Food GMP Manufactured Product | |
CAUTION: For Manufacturing, processing or repackaging | |
Bulk Pharmaceutical Chemical | |
Bulk Food Chemical | |
NF - Assay (HOCO(CHOH)₂COOH) (dried basis)(by acidimetry) | 99.7 - 100.5 % |
NF - Identification A | Passes Test |
NF - Identification B | Passes Test |
NF - Loss on Drying | ≤0.5 % |
NF - Residue on Ignition | ≤0.1 % |
NF - Limit of Oxalate | Passes Test |
NF - Sulfate (SO₄) | Passes Test |
NF - Optical Rotation [Å]²⁵^D (+) | 12.2 - 13.0 Degree |
FCC - Assay (HOCO(CHOH)₂COOH) (dried basis)(by acidimetry) | 99.7 - 100.5 % |
FCC - Identification A (IR) | Passes Test |
FCC - Identification B (Tartrate) | Passes Test |
FCC - Lead (Pb) | ≤2 mg/kg |
FCC - Limit of Oxalate | Passes Test |
FCC - Loss on Drying | ≤0.5 % |
FCC - Residue on Ignition | ≤0.05 % |
FCC - Specific (Optical) Rotation [Å]²⁵^D (+) | 12.2 - 13.0 Degree |
FCC - Sulfate (SO₄) | Passes Test |
Solubility (20 in 100) | Passes Test |
No Class 1,2,3 or other solvents are used or produced in the | |
manufacturing or purification of the product. | |
Elemental Impurities (USP 232, EP 5.20) - Information on elemental | |
impurities for this product is available on the associated Product | |
Regulatory Data Sheet and elemental impurity profile report. |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...