Triethylamine ≥99.5%, OR, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger

Synonyms: (Diethylamino)ethane, TEA

Management of Change (MOC) category = N

1961-59
MK196159CS 527.51 USD
MK196159
Triethylamine ≥99.5%, OR, Macron Fine Chemicals™
Triethylamine

Macron Fine Chemicals™ OR: organic reagents that are suitable for research applications

Formula: (C₂H₅)₃N
Boiling Pt: 90 °C (1013 hPa)
Melting Pt: –115 °C
Density: 0.729 g/cm³ (20 °C)
Flash Pt: –11 °C
Storage Temperature: Ambient
MDL Number: MFCD00009051
CAS Number: 121-44-8
EINECS: 204-469-4
UN: 1296
ADR: 3,II
Merck Index: 13,09740
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c2b1f184e438ecb01f88a4d90ee46c0d4316859d&vid=1c68408a0eedf1bfdb02d1b1e2337d50d449cf9d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Assay ((CH₃CH₂)₃N) ≥ 99.0 %
Color (APHA) ≤ 15
Water (H₂O) ≤ 1 %
Appearance (clear, colorless liquid free from foreign matter)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR