Tributyl phosphate, OR, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Warning

Synonyms: TBP, TBPA

Management of Change (MOC) category = N

1940-61
MK194061EA 248.93 USD
MK194061
Tributyl phosphate, OR, Macron Fine Chemicals™
Tributyl phosphate

Macron Fine Chemicals™ OR: organic reagents that are suitable for research applications

Formula: (CH₃(CH₂)₃O)₃PO
MW: 266.32 g/mol
Boiling Pt: 289 °C (1013 hPa)
Melting Pt: –79 °C
Density: 0.982 g/cm³ (20 °C)
Flash Pt: 151 °C
Storage Temperature: Ambient
MDL Number: MFCD00009436
CAS Number: 126-73-8
EINECS: 204-800-2
Merck Index: 13,09692
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fb9a0e9a8476470c1c50ab98587c559fec16cd8d&vid=6bcd8ac2f1665c53ba2c7ace4f34ebb3d890b549&prid=web&clid=SAPDAM

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Color (APHA) ≤50
Infrared Spectrum Passes Test
Appearance (clear, colorless liquid)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR