Benzyl alcohol ≥97%, AR®, Macron Fine Chemicals™
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
|
Warning
|
Management of Change (MOC) category = N
1816-04
MK181604CS
2209.2
USD
MK181604
Benzyl alcohol ≥97%, AR®, Macron Fine Chemicals™
Benzyl alcohol
Formula:
C₆H₅CH₂OH Boiling Pt: 205 °C (1013 hPa) Melting Pt: –15.3 °C Density: 1.05 g/cm³ (20 °C) Flash Pt: 101 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004599 CAS Number: 100-51-6 EINECS: 202-859-9 Merck Index: 13,01126 |
|
Specification Test Results
For Laboratory,Research,or Manufacturing Use | |
Assay (C₆H₅CH₂OH) | 97.0 - 100.5 % |
Residue on Ignition | ≤0.005 % |
Solubility | Passes Test |
Specific Gravity at 25°/25°C | 1.042 - 1.047 |
Identification | Passes Test |
Refractive Index, n²ºD | 1.539 - 1.541 |
Acidity | Passes Test |
Nonvolatile Residue | ≤0.05 % |
Halogenated Compounds and Halides (as Cl) | ≤0.03 % |
Benzaldehyde | ≤0.20 % |
Appearance (clear, colorless liquid) | |
The manufacturer has assurance, based on knowledge of the manufacturing process and controlled handling, shipping and storage of the material, that there is no potential for organic volatile impurities to be present as described in the current U.S.P. In this case unnecessary testing may be avoided. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...