Tannic acid ≥96.0% (dry basis) FCC, Macron Fine Chemicals™
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Management of Change (MOC) category = RL
MACR1674-21
MK1674-21EA
3187.51
USD
MK1674-21
Tannic acid ≥96.0% (dry basis) FCC, Macron Fine Chemicals™
Tannic acid
- Food GMP manufactured product
Formula:
C₇₆H₅₂O₄₆ Melting Pt: 218…219 °C Density: >1 g/cm³ (20 °C) Flash Pt: 199 °C |
MDL Number:
MFCD00066397 CAS Number: 1401-55-4 EINECS: 215-753-2 Merck Index: 12,09221 |
Specification Test Results
Food GMP Manufactured Product | |
Meets F.C.C. Requirements | |
CAUTION: For Manufacturing, processing or repackaging | |
Bulk Food Chemical | |
Assay () (dried basis) (FCC) | ≥96.0 % |
Appearance (fluffy, light tan to brown powder) | Passes Test |
Solubility | Passes Test |
Gums or Dextrin | Passes Test |
Resinous Substances | Passes Test |
Identification A | Passes Test |
Identification B | Passes Test |
Loss on Drying at 105°C | ≤7.0 % |
Residue on Ignition | ≤1.0 % |
Lead (Pb) | ≤2 mg/kg |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...