Sodium benzoate dust-free 99.0-101.0% (by anhydrous basis) NF, FCC, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Warning

Synonyms: Benzoic acid sodium salt

Management of Change (MOC) category = R

MACR1454-26
JT1454-26CS 2302.93 USD
JT1454-26 MK1454-26
Sodium benzoate dust-free 99.0-101.0% (by anhydrous basis) NF, FCC, Macron Fine Chemicals™
Sodium benzoate

GMP manufactured.

Formula: C₆H₅COONa
MW: 144.11 g/mol
Density: 1.44 g/cm³
MDL Number: MFCD00012463
CAS Number: 532-32-1

Order Now

Specification Test Results

Food GMP Manufactured Product
Meets F.C.C. Requirements
GMP Manufactured Product
Meets N.F. Requirements
CAUTION: For Manufacturing, processing or repackaging
Bulk Food Chemical
Bulk Pharmaceutical Chemical
NF - Assay (C₇H₅NaO₂) (anhydrous basis) 99.0 - 101.0 %
NF - Identification A Passes Test
NF - Identification B Passes Test
NF - Identification C Passes Test
NF - Alkalinity Passes Test
NF - Water (H₂O) ≤1.5 %
FCC - Assay (C₇H₅NaO₂) (anhydrous basis) 99.0 - 101.0 %
FCC - Arsenic (As) ≤3 ppm
FCC - Alkalinity (as NaOH) ≤0.04 %
FCC - Identification A Passes Test
FCC - Identification B Passes Test
FCC - Identification C Passes Test
FCC - Lead (Pb) ≤2 mg/kg
FCC - Water (H₂O) ≤1.5 %
Solubility Passes Test
Appearance (white, extruded cylinder) Passes Test
No Class 1,2,3 or other solvents are used or produced in the
manufacturing or purification of the product.
Elemental Impurities (USP 232, EP 5.20) - Information on elemental
impurities for this product is available on the associated Product
Regulatory Data Sheet and elemental impurity profile report.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR