Potassium iodide ≥99%, AR® ACS, free-flowing, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger

Synonyms: Dipotassium diiodide, Kali iodide

Management of Change (MOC) category = N

1123-12 1123-06 1123-20 1123-24 1123-02
MK112312CS 1512.7 USD
MK112312 MK112306 MK112320 MK112324 MK112302
Potassium iodide ≥99%, AR® ACS, free-flowing, Macron Fine Chemicals™
Potassium iodide

Granular.

Formula: Kl
Boiling Pt: 1330 °C (1013 hPa)
Melting Pt: 681 °C
Density: 3.13 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011405
CAS Number: 7681-11-0
EINECS: 231-659-4
Merck Index: 13,07727

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
ACS - Assay (KI) ≥99.0 %
ACS - Appearance (white, crystalline granules) Passes Test
ACS - Barium (Ba) ≤0.002 %
ACS - Calcium (Ca) ≤0.002 %
ACS - Chloride and Bromide (as Cl) ≤0.01 %
ACS - Heavy Metals (as Pb) ≤0.0005 %
ACS - Insoluble Matter ≤0.005 %
ACS - Iodate (IO₃) ≤3 ppm
ACS - Iron (Fe) ≤0.0003 %
ACS - Loss on Drying at 150°C ≤0.2 %
ACS - Magnesium (Mg) ≤0.001 %
ACS - pH of 5% Solution at 25°C 6.0 - 9.2
ACS - Phosphate (PO₄) ≤0.001 %
ACS - Sodium (Na) ≤0.005 %
ACS - Sulfate (SO₄) ≤0.005 %
Free Alkali, For Information Only
Total Alkali, For Information Only


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR