Bismuth(III) oxide ≥99.7%, AR®, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC

Synonyms: Bismuth Trioxide, Dibismuth trioxide

Management of Change (MOC) category = RL

MACR1057-24
JT1057-24CS 17468.51 USD
JT1057-24 MK1057-24
Bismuth(III) oxide ≥99.7%, AR®, Macron Fine Chemicals™
Bismuth (III) oxide
Formula: BiO₃
MW: 465.96 g/mol
Density: 8.90 g/cm³
MDL Number: MFCD00003462
CAS Number: 1304-76-3

Order Now

Specification Test Results

CAUTION: For Manufacturing, processing or repackaging
Assay (Bi₂O₃) ≥ 99.7 %
pH (by aqueous extraction) 6.75 - 9.25
Appearance (yellow powder) Passes Test
Arsenic (As) ≤ 0.0008 %
Chloride (Cl) ≤ 0.001 %
Copper (Cu) ≤ 0.01 %
Iron (Fe) ≤ 0.01 %
Sodium (Na) ≤ 0.03 %
On U.S. No. 325 Sieve ≤ 1.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR