tri-Sodium citrate dihydrate 99.0-100.5% (by anhydrous basis), crystalline, GenAR® USP, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC

Synonyms: Sodium Citrate Dihydrate, Sodium citrate tribasic dihydrate, Trisodium citrate, Sodium Citrate, Dihydrate, Sodium citrate dihydrate, Citric acid tridosium salt dihydrate, Sodium 2-hydroxypropane-1,2,3-tricarboxylate dihydrate, Citric acid trisodium salt dihydrate

Management of Change (MOC) category = R

MACR0634-45
JT0634-45CS 2352.1 USD
JT0634-45 MK0634-45
tri-Sodium citrate dihydrate 99.0-100.5% (by anhydrous basis), crystalline, GenAR® USP, Macron Fine Chemicals™
tri-Sodium citrate dihydrate

Caution: Not Intended for Parenteral Use

Formula: Na₃C₆H₅O₇·2H₂O
Melting Pt: 150 °C
Density: 1.70 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00150031
CAS Number: 6132-04-3
EINECS: 200-675-3
Merck Index: 13,08675
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e32147a215e8895f4b6db5aab1b99da667b795ac&vid=b929cda9d6f2eed18a8fefe57aa02b561af7c97f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

GMP Manufactured Product
Meets E.P. Chemical Specifications
Meets U.S.P Requirements
Meets J.P. Chemical Specifications
CAUTION: For Manufacturing, processing or repackaging
Bulk Pharmaceutical Chemical
USP - Assay (C₆H₅Na₃O₇) (anhydrous basis) 99.0 - 100.5 %
USP - Appearance (colorless or white to slightly yellow crystals) Passes Test
USP - Identification A Passes Test
USP - Identification B Passes Test
USP - Identification C Passes Test
USP - Alkalinity Passes Test
USP - Water (H₂O) 10.0 - 13.0 %
USP - Tartrate Passes Test
EP - Assay (anhydrous basis) 99.0 - 101.0 %
EP - Appearance of Solution (clear and colorless) Passes Test
EP - Identification A Passes Test
EP - Identification B Passes Test
EP - Acidity or Alkalinity Passes Test
EP - Readily Carbonizable Substances Passes Test
EP - Chlorides (Cl) ≤50 ppm
EP - Oxalate (C₂O₄) ≤300 ppm
EP - Sulfate (SO₄) ≤150 ppm
EP - Water (H₂O) 11.0 - 13.0 %
JP - Assay (anhydrous basis) 99.0 - 101.0 %
JP - Description Passes Test
JP - Identification A Passes Test
JP - Identification B Passes Test
JP - pH 7.5 - 8.5
JP - Clarity and Color of Solution Passes Test
JP - Chloride (Cl) ≤0.015 %
JP - Sulfate (SO₄) ≤0.048 %
JP - Heavy Metals ≤10 ppm
JP - Arsenic (As) ≤2 ppm
JP - Tartrate Passes Test
JP - Oxalate (C₂O₄) Passes Test
JP - Readily Carbonizable Substances Passes Test
JP - Loss on Drying at 180°C 10.0 - 13.0 %
Not Intended for Parenteral Use
No Class 1,2,3 or other solvents are used or produced in the
manufacturing or purification of the product.
Elemental Impurities (USP 232, EP 5.20) - Information on elemental
impurities for this product is available on the associated Product
Regulatory Data Sheet and elemental impurity profile report.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR