Citric acid monohydrate ≥99% (by titrimetric analysis), granules, AR® ACS, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Warning

Synonyms: 2-Hydroxypropane-1,2,3-tricarboxylic acid, Hydroxytricarballylic acid, 2-Hydroxy-1,2,3-propanetricarboxilic acid

Management of Change (MOC) category = N

0627-12 0627-20 0627-06
MK062712CS 581.16 USD
MK062712 MK062720 MK062706
Citric acid monohydrate ≥99% (by titrimetric analysis), granules, AR® ACS, Macron Fine Chemicals™
Citric acid monohydrate

Granular.

Formula: C₆H₈O₇.H₂O
Melting Pt: 135…152 °C
Density: 1.54 g/cm³ (20 °C)
Flash Pt: 173.9 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00149972
CAS Number: 5949-29-1
EINECS: 201-069-1
Merck Index: 13,02350
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3160681f96abd248f56672a96b72cc5f6e9d89a2&vid=c4db52c32c630d2c9876e60fc3e9e70a117ea0d8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
ACS - Assay (C₆H₈O₇·H₂O) (by titration) 99.0 - 102.0 %
ACS - Insoluble Matter ≤0.005 %
ACS - Residue after Ignition ≤0.02 %
ACS - Chloride (Cl) ≤0.001 %
ACS - Oxalate (C₂O₄) Passes Test
ACS - Phosphate (PO₄) ≤0.001 %
ACS - Sulfur Compounds (as SO₄) ≤0.002 %
ACS - Iron (Fe) ≤3 ppm
ACS - Lead (Pb) ≤2 ppm
ACS - Substances Carbonizable by Hot H₂SO₄ Passes Test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR