Citric acid monohydrate ≥99.5% (by anhydrous basis), granules USP, Macron Fine Chemicals™
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
|
Warning
|
Synonyms:
2-Hydroxypropane-1,2,3-tricarboxylic acid, Hydroxytricarballylic acid, 2-Hydroxy-1,2,3-propanetricarboxilic acid
Management of Change (MOC) category = R
0616-12
0616-88
MACR0616-06
MK061612CS
362.24
USD
MK061612
MK061688
MK0616-06
JT0616-26
Citric acid monohydrate ≥99.5% (by anhydrous basis), granules USP, Macron Fine Chemicals™
Citric acid monohydrate
Granular. GMP manufactured.
Caution: Must be subjected to further processing during the preparation of injectable dosage forms.
Formula:
C₆H₈O₇.H₂O Melting Pt: 135…152 °C Density: 1.552 g/cm³ (20 °C) Flash Pt: 173.9 °C (closed cup) Storage Temperature: Ambient |
MDL Number:
MFCD00149972 CAS Number: 5949-29-1 EINECS: 201-069-1 Merck Index: 13,02350 |
|
Specification Test Results
Meets U.S.P Requirements | |
GMP Manufactured Product | |
Preserve in Tight Containers | |
CAUTION: For Manufacturing, processing or repackaging | |
Bulk Pharmaceutical Chemical | |
USP - Assay (as C₆H₈O₇) (calculated on anhydrous basis) | 99.5 - 100.5 % |
USP - Clarity of Solution | Passes Test |
USP - Color of Solution | Passes Test |
USP - Identification | Passes Test |
USP - Water (H₂O)(by Karl Fischer titrn) | 7.5 - 9.0 % |
USP - Residue on Ignition | ≤0.1 % |
USP - Readily Carbonizable Substances | Passes Test |
USP - Sulfate (SO₄) | ≤0.015 % |
USP - Limit of Oxalic Acid | ≤0.036 % |
Solubility (5 in 50) | Passes Test |
No Class 1,2,3 or other solvents are used or produced in the | |
manufacturing or purification of the product. | |
Must be subjected to further processing during the preparation of | |
injectable dosage forms. | |
Elemental Impurities (USP 232, EP 5.20) - Information on elemental | |
impurities for this product is available on the associated Product | |
Regulatory Data Sheet and elemental impurity profile report. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...