Zinc phenolsulfonate, granular, purified, Macron Fine Chemicals™
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
|
Warning
|
Management of Change (MOC) category = NI
MACR0083-24
MACR0083-41
MK0083-24EA
7508.4
USD
MK0083-24
MK0083-41
Zinc phenolsulfonate, granular, purified, Macron Fine Chemicals™
Zinc phenolsulfonate
Formula:
Zn(C₆H₅O₄S)₂ Melting Pt: 257 °C |
CAS Number:
127-82-2 |
Specification Test Results
For Laboratory,Research,or Manufacturing Use | |
Assay (Zn(C₆H₅O₄S)₂) | 73.7 - 77.4 % |
Appearance | Passes Test |
Identification A | Passes Test |
Identification B | Passes Test |
Identification C | Passes Test |
Sulfate (SO₄) | ≤0.04 % |
Solubility (clear, colorless solution) | Passes Test |
Arsenic (As) | ≤0.0003 % |
Lead (Pb) | ≤0.0020 % |
pH of 0.4% Solution at 25°C, For Information Only | |
Iron (Fe) | ≤0.0005 % |
Solubility in Alcohol (NTU) | ≤10 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...