Guanidinium hydrochloride ≥99.5%, NF-GenAR® for biotechnology, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Warning


Management of Change (MOC) category = R

MACR0072-26
MK007220EA 6732.04 USD
MK007220 MK0072-26
Guanidinium hydrochloride ≥99.5%, NF-GenAR® for biotechnology, Macron Fine Chemicals™
Guanidinium hydrochloride
Formula: NH₂C(=NH)NH₂·HCl
Melting Pt: 185 °C
Density: 1.354 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00013026
CAS Number: 50-01-1
EINECS: 200-002-3
Merck Index: 13,04578
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=da5a871cab977e6d9dc666b457732eb1e869a714&vid=b0d4fcb044b6106fce8e40448dbf2ce6e7077ad7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

GMP Manufactured Product
CAUTION: For Manufacturing, processing or repackaging
Assay (NH₂C(:NH)NH₂·HCl) ≥99.5 %
Acidity (as HCl) ≤0.01 %
Arsenic and Antimony (as As) ≤10 ppm
Copper (Cu) ≤10 ppm
Heavy Metals (as Pb) ≤10 ppm
Iron (Fe) ≤10 ppm
Loss on Drying at 105°C ≤0.5 %
Enzyme Activity - DNase Activity Passes Test
Enzyme Activity - RNase Activity Passes Test
Enzyme Activity - Protease Activity Passes Test
Melamine and Related Compounds Passes Test
Absorbance of a 6M Solution (260 nm) (au) ≤0.030
Residue after Ignition ≤0.20 %
Water-Insoluble Matter ≤0.3 %
Product Information (not a specification): Appearance - colorless
crystals
MANUFACTURED EXCLUSIVELY FOR AVANTOR PERFORMANCE MATERIALS, LLC., BY
BIOSPECTRA, INC., STROUDSBURG, PA


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR