Acetonitrile ≥99.5% (by GC, corrected for water content), AR® ACS, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger


Management of Change (MOC) category = N

0043-08 0043-06 MACR0043-26
MK004308CS 2635.84 USD
MK004308 MK004306 MK0043-26
Acetonitrile ≥99.5% (by GC, corrected for water content), AR® ACS, Macron Fine Chemicals™
Acetonitrile
Formula: H₃CCN
Boiling Pt: 81.6 °C (1013 hPa)
Melting Pt: –45.7 °C
Density: 0.79 g/cm³
Flash Pt: 2 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00001878
CAS Number: 75-05-8
EINECS: 200-835-2
UN: 1648
ADR: 3,II
REACH: 01-2119471307-38
Merck Index: 13,00071
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cef8be50fcca82050c5bda827ad9c5f57f4c5e27&vid=b80b5c39e654fcf6904b18b58e22a816e8e138b7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
ACS - Assay (CH₃CN) (by GC, corrected for water) ≥99.5 %
ACS - Appearance (clear, colorless liquid, free of extraneous matter) Passes Test
ACS - Color (APHA) ≤10
ACS - Residue after Evaporation ≤0.005 %
ACS - Titrable Acid (µeq/g) ≤8
ACS - Titrable Base (µeq/g) ≤0.6
ACS - Water (H₂O)(by Karl Fischer titrn) ≤0.3 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR