Monoject™ Curved Tip Syringe, Covidien
Supplier: Cardinal Health
8881412012
MJ8881-412012BX 81.42 USD
MJ8881-412012
Monoject™ Curved Tip Syringe, Covidien
Syringes Syringes, Medical
This Monoject™ specialty syringe is ungraduated and features a 12mL capacity
Equipped with a reusable plastic finger grip extender, it is ideal for pressure irrigation.