Two-Way Pilot-Operated Valves with Manual Override
Supplier: BURKERT FLUID CONTROL SYSTEMS
453349M MFLX98604-10 MFLX98604-20
MFLX98604-00EA 1010 USD
MFLX98604-00 MFLX98604-10 MFLX98604-20
Two-Way Pilot-Operated Valves with Manual Override
Valves Solenoid Valves
Use under full vacuum.
  • Constructed of PVC body and EPDM diaphragm
Two-Way Pilot-Operated Valves with Manual Override (Normally closed; require 7 psi differential pressure. Require 32-mm DIN plug)