Two-Way Direct-Lift Solenoid Valves
Supplier: VWR International
MFLX98554-84 MFLX98554-62 MFLX98554-82
MFLX98554-84EA 1080 USD
MFLX98554-84 MFLX98554-62 MFLX98554-82
Two-Way Direct-Lift Solenoid Valves
Valves Solenoid Valves
Use under full vacuum.
  • Chemical-resistant for aggressive fluids
  • Voltage tolerance is ±10%
  • 100% duty cycle
Two-way direct lift valves are normally closed and feature bellows made of PTFE and seals made of Viton®.
Choose from valves with a PVC, polypropylene, or PVDF body for a wide range of agreesive fluid applications.