Masterflex® Ismatec® Fixed-Speed Rack-Mount Minicartridge Peristaltic Pumps, Avantor®
Supplier: Avantor Fluid Handling
ISM847B-A230V50 ISM848-230V50H ISM849-230V50H ISM852-230V50H ISM853-230V50H ISM850A-230V50H ISM851-230V50H ISM846-230V50H ISM844-230V50H ISM845-230V50H ISM847B-A115V60 ISM849-115V60H ISM852-115V60H ISM853-115V60H ISM850-115V60H ISM851-115V60H ISM846-115V60H ISM844-115V60H ISM845-115V60H
MFLX78012-05CS 1030 USD
MFLX78012-05 MFLX78012-15 MFLX78012-25 MFLX78012-35 MFLX78012-45 MFLX78012-55 MFLX78012-65 MFLX78012-85 MFLX78012-95 MFLX78012-97 MFLX78012-00 MFLX78012-20 MFLX78012-30 MFLX78012-40 MFLX78012-50 MFLX78012-60 MFLX78012-80 MFLX78012-90 MFLX78012-92
Masterflex® Ismatec® Fixed-Speed Rack-Mount Minicartridge Peristaltic Pumps, Avantor®
Pumps Peristaltic Pumps
Compact size mounting rod—use these pumps almost anywhere.
  • A constant-speed, synchronous motor ensures a steady, repeatable flow rate
  • Change tubing sizes to obtain different flow rates
Minicartridge design offers the easiest and most convenient tubing changes of all—even replace tubing while your pump is running! Order extra optional minicartridges with adjustable occlusion to manually vary pressure on each channel. Have extra 3-stop tubing on hand when fast tubing changes are crucial. (Scroll down to see all accessory choices.)
Delivery information: Includes a 5-1/2"L x 1/2" dia (14 x 0.5 cm) mounting rod, a 6-ft (1.8-m) power cord with plug.