Masterflex® I/P® Digital Modular Drive Systems, Avantor®
Supplier: Avantor Fluid Handling
77962-20 77962-30
MFLX77962-20EA 5550 USD
MFLX77962-20 MFLX77962-30
Masterflex® I/P® Digital Modular Drive Systems, Avantor®
Pumps Peristaltic Pumps
Place motor and controller where convenient with modular design.
  • Flow rate 0.06 to 8 LPM with I/P tubing—flow rate depends on drive rpm, pump head, and tubing size
  • Digital interface with LED display shows pump rpm— confirm speed setting at a glance
  • Remote control available using analog communication—voltage, current and contact closure
  • Powerful motor lets you stack two pump heads for increased flow rate while offering ±0.25% speed control accuracy
  • Membrane keypad allows for easy up-front control of speed, direction, and start/stop
I/P® modular drives feature reversible pumping while allowing you the flexibility to separate the motor from the controller. These accurate modular drives have powerful motors, speed display, and ±0.25% speed control for precise flow regulation—ideal for metering and critical transfer applications.