Masterflex® L/S® Standard Digital PTFE-Tubing Pump System, Avantor®
Supplier: Avantor Fluid Handling
77912-10
MFLX77912-10EA 3480 USD
MFLX77912-10
Masterflex® L/S® Standard Digital PTFE-Tubing Pump System, Avantor®
Pumps Peristaltic Pumps
Transfer aggressive organic solvents and high-purity fluids.
  • Flow range 0.18 to 65 ml/min—low rate depends on drive rpm and 4- or 6-mm OD tubing
  • Rigid pump tubing designed to pump fluids at pressures up to 100 psi (6.9 bar)
  • Adjustable occlusion—tighten for higher pressure, loosen for longer tubing life
  • Limited chemical exposure with PTFE tubing as only wetted part
This complete Masterflex system is ideal for filtration, chemical injection, and other high pressure applications. The PTFE pump head accepts either 4- or 6-mm OD rigid PTFE-tubing sets. Note: Use Masterflex® PTFE-tubing sets only.
Delivery information: Includes l/S® PTFE-tubing pump head 77390-00, PTFE-tubing set 77390-60, and L/S® variable-speed drive 07528-20 (3 to 300 rpm). System also includes two 1/4" NPT(M) pipe adapters.