Masterflex® Bioprocess Pump Cart
Supplier: Avantor Fluid Handling B.V.
77112-90
MFLX77112-90EA 4600 USD
MFLX77112-90
Masterflex® Bioprocess Pump Cart
Carts
This 304 stainless steel cart is designed specifically for MasterSense™ B/T® and I/P® Process pumps for use in cleanroom environments. The cart features locking castors, tubing and cord wrap holder and includes mounting hardware for Masterflex® MasterSense™ B/T® pump systems.
  • Designed for portability and operation with the pump mounted securely to the platform cart
  • 304 SS design allows for compatibility with common cleaning solutions
  • <32 Ra surface polish on all contact surfaces for easy cleanability
  • Mounting pattern supports various configurations
  • Rounded surfaces - no sharp edges
  • Locking castors prevent movement of pump system while in service
  • Ergonomic hand rail and light weight allows for greater maneuverability
  • Cord wrap serves dual function of securing pump tubing and/or power cable during transport
Designed for easy and secure operation of Masterflex pump systems where on-demand pumping and portability is required. Ideal for use where flexible drug manufacturing is used, cart easily allows relocation of pump to best location for efficient operation.
Certifications: ROHS, REACH, ISO9001, CE.
Accessories information: Pumps not included. Order mounting plates for use with I/P® and L/S® series MasterSense™ pumps separately (not required for single B/T® pump mounting).
Delivery information: Includes cart and mounting hardware for a single B/T pump system (order separately 77112-10).
Packaging: Cart ships on pallet.