Masterflex® Liquid Detector Pad Cables, Avantor®
Supplier: Avantor Fluid Handling
77095-02 MFLX77095-00
MFLX77095-02CS 77 USD
MFLX77095-02 MFLX77095-00
Masterflex® Liquid Detector Pad Cables, Avantor®
Cables
Make the most of your Liqui-Sense leak detector with suitable replacement parts.
  • Compatible with Masterflex® Liqui-Sense controllers
This replacement cable features a 6-pin cable with RJ-12 connections and has a 3.5 foot cable length.