Masterflex® Single-Head Solenoid Diaphragm Pumps, High-Volume, Avantor®
Supplier: VWR International
C15N443X000V00
MFLX76302-40EA 709 USD
MFLX76302-40 MFLX07135-57
Masterflex® Single-Head Solenoid Diaphragm Pumps, High-Volume, Avantor®
Pumps Diaphragm Pumps
Economical, chemically-compatible diaphragm metering pumps.
  • Flow rates from 662 to 2081 ml/min
  • Overload-protected, fan-cooled, shaded-pole gear motor (1/35 hp) is permanently lubricated
  • Pump greater flow rates at lower pressures
  • Feature an easily operated cam-type adjustment with an infinite number of settings bet
Greater flow rates are pumped at lower pressures. Feature an easily operated cam-type adjustment with an infinite number of settings between zero and 100% of maximum pump capacity.
Delivery information: Includes foot valve/strainer, polypropylene antisiphon valve, vinyl inlet and outlet tubing, mounting hardware, and 6-ft, three-wire cord and plug.