Masterflex® Programmable Compact Syringe Pump, Avantor®
Supplier: VWR International
MFLX75900-00
MFLX75900-00EA 1140 USD
MFLX75900-00
Masterflex® Programmable Compact Syringe Pump, Avantor®
Pumps Syringe Pumps
Economical OEM-style pump.
  • Flow rates from 0.73 uL/hr to 1699 ml/hr
  • Download program from a computer to the pump to operate the pump independently
  • Accessories allow networking of up to 100 pumps
Pre-program the pump with a dispensing program, then operate the pump via an attached footswitch, button, or I/O controller. Dispensing software allows user to infuse and withdraw in uL/hr, uL/min, ml/hr, and ml/min. Design a program to change direction
Power supply is not included.
For the US Power Supply, please look at 75900-93.
For the EU Power Supply, please look at 75900-94.