Masterflex® Submersible Sampling Pumps, Avantor®
Supplier: VWR International
75509-60
MFLX75509-55EA 331.7 USD
MFLX75509-55 MFLX75509-65 MFLX75509-50
Masterflex® Submersible Sampling Pumps, Avantor®
Pumps Centrifugal Pumps
12 VDC powered is ideal for those field well purging/sampling applications.
  • Practical for dedicated use and disposal
These Masterflex® submersible sampling pumps are suitable for purging and sampling applications. Attach pump to 1/2” or 3/8” tubing (tubing sold separately), and lower pump directly into well. Pumps are constructed with an ABS plastic and polycarbonate body, stainless steel impeller, and nitrile seal.