Masterflex® High-Performance Dual-Piston Pumps, Avantor®
Supplier: VWR International
HF300SRX
MFLX74930-01EA 31960 USD
MFLX74930-01
Masterflex® High-Performance Dual-Piston Pumps, Avantor®
Pumps Piston Pumps
Dual-head design for precise flow rates and low pulsation.
  • Flow rates from 0.1 to 250.0 ml/min
  • Pump features a servo motor, LCD keypad, and RS-232 interface
  • Pressure is 9000 psi (612 bar) from 0 to 210 ml/min or 6500 psi from 211 to 250 ml/min
  • Flow precision of 0.5% RSD and an accuracy of ≤2% of set point
Thoughtful design is shown through the whole pump—from the dual pump heads allowing for very accurate flow rates and low pulsation to the readily accessible check valves, pistons and seals making for fast and easy maintenance.
Standard configuration is w