Masterflex® Dual, Independent Channel, Touch-Screen Syringe Pump, Avantor®
Supplier: VWR International
788088C
MFLX74906-05EA 7690 USD
MFLX74906-05
Masterflex® Dual, Independent Channel, Touch-Screen Syringe Pump, Avantor®
Pumps Syringe Pumps
No need for two separate pumps.
  • Flow rates as low as 1.02 pL/min
  • Intuitive graphical interface with large 7" color LCD touch screen control is easy to use
  • Daisy chain and remote control compatible (order cables separately)
  • Syringe sizes accepted: 0.5 µL to 60 mL
This dual-channel syringe pump functions like two separate pumps. Each channel has its own motor and plunger drive, so you can program different syringe sizes, dispensing rates, and alternate directions. Large touch screen and user-friendly icon-driven so
Delivery information: Includes power supply.