Pulsafeeder 4 to 20 mA Pace Pumps
Supplier: EQUIP SOLUTIONS
LPH8MA-WTCB-XXX
MFLX74141-60EA 2930 USD
MFLX74141-60 MFLX74141-00 MFLX74141-10 MFLX74141-20 MFLX74141-30 MFLX74141-40 MFLX74141-50
Pulsafeeder 4 to 20 mA Pace Pumps
Pumps Diaphragm Pumps
Reliable metering performance—guided dual check valves provide precise seating and excellent priming and suction lift.
  • PVC and PTFE construction with external control
The 4 to 20 mA pumps feature automatic control with 4 to 20 mADC direct or external pacing, reliable timing circuits, stop function, and manually adjustable stroke rate and length. Operate pump in automatic or manual mode by selecting the proper setting on the "Auto-Off-Manual" switch. Circuit protection guards against voltage and current upsets. Install pumps indoors or outdoors.