Masterflex® Analog Gear Pump Systems, Avantor®
Supplier: Avantor Fluid Handling
74013-02 75211-17 & 07002-14 75211-12 & 74012-11 74013-22 74013-32 74013-42 75211-17 & 07003-04 75211-17 & 74012-51 74013-72 75211-17 & 74012-11 74013-37 75211-12 & 74012-51 74013-77 75211-17 & 74012-21 74013-77-AU 74013-77-DK 74013-77-IN 74013-77-EU 74013-77-CH 74013-77-UK 74013-77-IT 74013-77-IL
MFLX74013-02EA 2480 USD
MFLX74013-02 MFLX74013-07 MFLX74013-12 MFLX74013-22 MFLX74013-32 MFLX74013-42 MFLX74013-47 MFLX74013-57 MFLX74013-72 MFLX74013-17 MFLX74013-37 MFLX74013-52 MFLX74013-77 MFLX74013-27 MFLX74013-77-AU MFLX74013-77-DK MFLX74013-77-IN MFLX74013-77-USS MFLX74013-77-EU MFLX74013-77-CH MFLX74013-77-UK MFLX74013-77-IT MFLX74013-77-USN MFLX74013-77-IL
Masterflex® Analog Gear Pump Systems, Avantor®
Pumps Gear Pumps
Simple variable speed control handles many fluid handling applications.
  • 100:1 speed-control range controls flow over a wide range
  • Simple operation with single-turn speed control
  • Separate power switch enables you to maintain speed setting when turning drive on/off; front-panel LED indicates power on
  • Remote control of start/stop via DB9 connector on back of drive
Compact, cost-effective drive offers simple variable speed control for many fluid handling applications from general transfer to filtration to chemical injection. Conveniently coupled with an A-mount gear pump head for pulseless fluid flow.