Micropump Cavity-Style Gear Pump Heads for NEMA Type 56 C-face Motors
Supplier: MICROPUMP INC
L20563+83400 MFLX73007-00 MFLX73007-11
MFLX73007-02EA 5230 USD
MFLX73007-02 MFLX73007-00 MFLX73007-11
Micropump Cavity-Style Gear Pump Heads for NEMA Type 56 C-face Motors
Pumps Pump Heads Gear Pump Heads
Smooth pulseless flow provides precise, leak-free delivery of fluids.
  • Compact size and easy to service
  • Cavity-style heads are designed to handle higher inlet pressures.
Magnetically driven, precision-geared cavity pump heads provide smooth, accurate, pulseless fluid delivery. The magnetic drive and a single seal keep fluids securely inside and potential contaminants out. Chemical resistant materials handle a wide range of fluids.