Masterflex® Acid/Base Metering Pump Systems, Avantor®
Supplier: VWR International
MFLX70790-40
MFLX70790-40EA 6790 USD
MFLX70790-40
Masterflex® Acid/Base Metering Pump Systems, Avantor®
Pumps Diaphragm Pumps
Complete system—all in one.
  • Maximum flow range of 6 to 48 GPD│Controller with two programmable rely outputs
Controller has two programmable relay outputs and separate alarm relay. Large dual LCD shows pH (or mV) and temperature simultaneously. Continuous-duty pump features an adjustable stroke rate and length, reliable timing circuits, and leak-free bleed valves for safe and easy priming. The pH probe is a self-cleaning, double-junction electrode that prevents suspended solids build-up on electrode surface. Injection quill injects corrosive chemicals in the center of the pipe — preventing damage to the pipe wall.
Please contact your local Avantor office to customize a system suited to your needs.
Delivery information: Includes eutech® pH 2000 controller, Pulsafeeder® metering pump, plumbing connections, submersible pH probe, PVC injection quill, and one pump service kit.