Chemsteel Gear Pumps with Mechanical Seals
Supplier: VWR International
OBS21416CA-T97
MFLX70736-00EA 2921 USD
MFLX70736-00 MFLX70736-05 MFLX70736-60 MFLX70736-62 MFLX70736-68 MFLX70736-72 MFLX70736-74
Chemsteel Gear Pumps with Mechanical Seals
Pumps Gear Pumps
Self-priming and bidirectional capabilities offer excellent dosing, metering, and transfer of polymers and corrosive chemicals.
  • Wear-resistant helical gears offer reduced noise levels of 10 dB
  • Heavy-duty three-piece design combines bearings and wear plates to easily service pump in the field
316 SS Gear Pumps with mechanical seals provide a cost-effective option for pumping a variety of fluids.
All pumps are close-coupled to standard 56C-face motors. Motor type is TEFC, totally enclosed, fan-cooled. The 115/230 VAC models are single phase. Three phase motors for variable-speed use with controllers are available. Call our Application Specialists to order.
Also contact our Application Specialists if your maximum viscosity requirements are higher.
Service kits include: drive and idler gears mounted to the shafts, keys, bearings, O-rings, and seal assemblies.