Hand-Operated FDA Compliant Drum Pumps
Supplier: BECKSON INDUSTRIAL PRODUCTS
CF1209S MFLX70607-22
MFLX70607-20EA 106 USD
MFLX70607-20 MFLX70607-22
Hand-Operated FDA Compliant Drum Pumps
Pumps Drum/Transfer Pumps
Ideal for most acid/base or ionic salt applications.
  • Materials are FDA compliant for indirect food contact
  • Available with outlet hose (70607-20) or spout (70607-22)
These plastic hand-operated drum pumps are constructed with materials that conform under FDA CFR 21 regulations for indirect food contact. Pumps features 26 strokes/gallon (7 strokes/liter) with 3/4” NPT(M) container adapter. Both pumps are constructed with PVC barrel, Viton® valves and seals, and LDPE tube. Choose between models with a PVC hose outlet or LDPE spout outlet.