Flux® Drum Pump Tube Sets, PVDF
Supplier: FLUX PUMPS CORP
F424PVDF41/36-47" F424PVDF41/36-39" MFLX70601-00 MFLX70601-24
MFLX70601-04EA 1410 USD
MFLX70601-04 MFLX70601-02 MFLX70601-00 MFLX70601-24
Flux® Drum Pump Tube Sets, PVDF
Pumps Drum/Transfer Pumps
Construct a pump system that meets your specific needs.
  • PVDF sets are ideal for your high-purity applications
Standard, High-Head, and Heavy-Duty Tube Sets feature a sealless design for low maintenance and long life. All tube sets include a hose barb fitting made of the same material as the tube set. Fitting accepts either 3/4" or 1" ID tubing.