Battery-Powered Drum Pumps, Polypropylene
Supplier: BPH PUMP & EQUIPMENT INC
0205-055-1
MFLX70510-42EA 1480 USD
MFLX70510-42 MFLX70510-44 MFLX70510-40
Battery-Powered Drum Pumps, Polypropylene
Pumps Drum/Transfer Pumps
Battery power provides the greatest flexibility in your lab.
  • Flows up to 4.2 GPM (16 LPM) or 26.2 ft (8 m) of head resistance
  • Variable flow control is achieved with easy-to-use dial
  • Three different sizes allow for simple choices based upon container size
  • Battery charger accommodates both 120 VAC, 60 Hz and 220 VAC, 50 Hz
Do not run dry.
Power is from 9.6 V NiMH internal battery with charger (120 VAC, 60 Hz and 220 VAC, 50 Hz)