Masterflex® Correlated and Direct-Reading Variable Area Flowtubes for Gases, Avantor®
Supplier: AALBORG INSTRUMENTS & CONTROL
023-05G 365-17ST 014-18G 014-19ST 042-09ST 032-20G 022-24S 022-01C N042-01ST N014-16ST N032-16ST 032-08S N022-15G N013-25ST N042-11S N062-13G N062-07C N092-05G N062-12G N112-06S 062-02ST MFLX32048-03 MFLX32048-04 MFLX32048-10 MFLX32048-11 MFLX32048-14 MFLX32048-15 MFLX32048-16 MFLX32048-18 MFLX32048-19 MFLX32048-21 MFLX32048-25 MFLX32048-26 MFLX32048-28 MFLX32048-29 MFLX32048-30 MFLX32048-31 MFLX32048-32 MFLX32048-50 MFLX32048-53 MFLX32048-57 MFLX32048-58 MFLX32048-61 MFLX32048-62 MFLX32048-64 MFLX32048-65 MFLX32048-66 MFLX32048-68
MFLX32048-00EA 57.2 USD
MFLX32048-00 MFLX32048-01 MFLX32048-05 MFLX32048-06 MFLX32048-07 MFLX32048-08 MFLX32048-09 MFLX32048-12 MFLX32048-17 MFLX32048-20 MFLX32048-22 MFLX32048-23 MFLX32048-24 MFLX32048-27 MFLX32048-51 MFLX32048-55 MFLX32048-56 MFLX32048-59 MFLX32048-60 MFLX32048-63 MFLX32048-67 MFLX32048-03 MFLX32048-04 MFLX32048-10 MFLX32048-11 MFLX32048-14 MFLX32048-15 MFLX32048-16 MFLX32048-18 MFLX32048-19 MFLX32048-21 MFLX32048-25 MFLX32048-26 MFLX32048-28 MFLX32048-29 MFLX32048-30 MFLX32048-31 MFLX32048-32 MFLX32048-50 MFLX32048-53 MFLX32048-57 MFLX32048-58 MFLX32048-61 MFLX32048-62 MFLX32048-64 MFLX32048-65 MFLX32048-66 MFLX32048-68
Masterflex® Correlated and Direct-Reading Variable Area Flowtubes for Gases, Avantor®
Flow Meters
Create your own flowmeter systems for complete flexibility.
  • Correlated flowmeters feature a generic scale—reference flow curves calibrated to a variety of media, temperatures and pressures to get the flow rate
  • Direct-reading flowmeters feature a calibrated scale for a specific medium—no need to convert readings to get flow rate
  • Use with air or water for laboratory and industrial applications
  • Required system components for flowmeter systems would be frame, flowtube(s), valve cartridge, and tripod—order each component separately
  • Minimum flow rate is approximately 10% of max flow rate
Design a flow system to meet your metering, mixing, or monitoring applications. We offer a variety of frames and flowtubes so that you can create the system to meet your exact needs.
Choose 65- or 150-mm flowtubes depending on the selected frame size. Flowtubes are calibrated for direct-reading or correlated reading of air or water at STP (standard temperature and pressure).