Smiths Medical® Large-Bore Stopcock Fittings, Male Luer Lock
Supplier: ICU MEDICAL, INC.
B19311LG2 (1000PK)
MFLX30600-23BLKEA 944 USD
MFLX30600-23BLK
Smiths Medical® Large-Bore Stopcock Fittings, Male Luer Lock
Valves Ball Valves
Enhance fluid flow with a larger bore than standard luer stopcocks.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
Large-bore adapters are perfect for handling higher flow volumes in process while using connections similar to luer systems.