Slim-Line Double-Open-End Filter Cartridge Housings
Supplier: PENTAIR RESIDENTIAL FILTRATION LLC
158005 MFLX01508-31 MFLX29820-30 MFLX29820-36 MFLX29820-37
MFLX29820-00EA 23.2 USD
MFLX29820-00 MFLX29820-01 MFLX29820-10 MFLX29820-11 MFLX29820-32 MFLX29820-33 MFLX29820-34 MFLX29820-35 MFLX01508-31 MFLX29820-30 MFLX29820-36 MFLX29820-37
Slim-Line Double-Open-End Filter Cartridge Housings
Filters Filter Cartridges
Visually examine cartridge with clear housing.
  • Choose between Styrene-Acrylonitrile (SAN) or polypropylene (PP) housing construction
Slim-Line Housing with Styrene-Acrylonitrile (SAN) has been stress-relieved for added clarity and strength. Clear body allows on site examination of flow and cartridge performance. Housing has 3/8" connections for medium to light duty applications. Maximum pressure is 125 psi, maximum temperature is 125 °F (51.7 °C). Buna-N seal.