Accessories for Cartridge Filter Housings
Supplier: PENTAIR RESIDENTIAL FILTRATION LLC
MFLX01508-38 150295 MFLX01508-39 MFLX01508-47 MFLX01509-38
MFLX01508-38EA 24.2 USD
MFLX01508-38 MFLX01508-46 MFLX29802-52 MFLX29802-54 MFLX01508-39 MFLX01508-47 MFLX01509-38
Accessories for Cartridge Filter Housings
Filters Filter Cartridges
Optimize performance of your cartridge filter housing system.
  • Choose from a wide range of replacement parts and accessories
Spanner wrench loosens the filter housing cap to make filter replacement easier.