Big Blue Filter Cartridge Housings, Polypropylene
Supplier: PENTAIR RESIDENTIAL FILTRATION LLC
150235 MFLX01507-77 MFLX29802-01 MFLX29802-10
MFLX01507-76EA 74.3 USD
MFLX01507-76 MFLX01507-78 MFLX01508-65 MFLX01508-67 MFLX01508-70 MFLX01508-71 MFLX01508-75 MFLX29802-00 MFLX29802-11 MFLX01507-77 MFLX29802-01 MFLX29802-10
Big Blue Filter Cartridge Housings, Polypropylene
Filters Filter Cartridges
Flow rates up to 50 GPM in a compact, relatively low-cost package.
  • Constructed of durable Polypropylene
These systems offer up to four times the filtration area than standard cartridge systems, which makes them ideal for high-flow processes. A wide range of cartridges is offered for numerous applications. Order housings and cartridges separately.
Housing and cap are constructed of durable polypropylene and sealed with a Buna-N O-ring. Housing is suitable to 100 °F (37 °C); maximum operating pressure is 100 psi (6.9 bar).