Masterflex® L/S® Tilt Bail for Digital Drives, Avantor®
Supplier: Avantor Fluid Handling
7523-98
MFLX07523-98EA 174 USD
MFLX07523-98
Masterflex® L/S® Tilt Bail for Digital Drives, Avantor®
Mounting Plates
Use to securely elevate front-end of digital drive.
  • Constructed with steel-plating for enhanced durability and corrosion resistance
Steel-plated elevating foot resists corrosion and comes with everything needed to mount onto your digital drive. Simple design elevates the front of the drive and swings out of the way when not in use.
Compatible with the following drives:
07522-20, 07552-30
07523-80, 07523-90
07528-10, 07528-20, 07528-30
07551-00, 07551-10
Compatible with the controller of the following modular drive:
77923-80